APO455826727 Apollo Scientific Ltd

2-(3-Bromophenyl)-1-(pyrrolidin-1-yl)ethan-1-one

  • MDL number MFCD09800919
  • Cas Number : 951884-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[(3-Bromophenyl)acetyl]pyrrolidine, 1-Bromo-3-[2-oxo-2-(pyrrolidin-1-yl)ethyl]benzene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=41293

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=41293
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart