APO455826728 Apollo Scientific Ltd

7-Methoxyimidazo[1,2-a]pyridine-8-carbonitrile

  • MDL number MFCD09801017
  • Cas Number : 834869-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-218
Synonym 8-Cyano-7-methoxyimidazo[1,2-a]pyridine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=41301

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=41301
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart