APO455826729 Apollo Scientific Ltd

5-Bromo-2-hydrazino-3-nitropyridine

  • MDL number MFCD09801053
  • Cas Number : 15862-42-7
  • Molecular Weight : 233.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-hydrazino-3-nitropyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-hydrazino-3-nitropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart