APO455826732 Apollo Scientific Ltd

4-(tert-Butylamino)-6-chloropyrimidine

  • MDL number MFCD09756959
  • Cas Number : 945896-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-(tert-Butyl)-6-chloropyrimidin-4-amine, 4-(tert-Butylamino)-6-chloro-1,3-diazine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=41324

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=41324
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart