APO455826757 Apollo Scientific Ltd

2-Bromoethyl 4-(tert-butyl)phenyl ether

  • MDL number MFCD02030708
  • Cas Number : 5952-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 95/0.38mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37094

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37094
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart