APO455826762 Apollo Scientific Ltd

6-Bromoquinazoline-2,4(1H,3H)-dione

  • MDL number MFCD00462868
  • Cas Number : 88145-89-5
  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 6-Bromo-2,4-dioxo-1,2,3,4-tetrahydroquinazoline
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromoquinazoline-241h3h-dione

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromoquinazoline-241h3h-dione
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart