APO455826764 Apollo Scientific Ltd

5-Chloro-2-hydrazinopyridine

  • MDL number MFCD07786313
  • Cas Number : 27032-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-128
Synonym (5-Chloropyridin-2-yl)hydrazine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-hydrazinopyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-hydrazinopyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart