APO455826768 Apollo Scientific Ltd

trans-2,5-Dichlorocinnamic acid

  • MDL number MFCD03002786
  • Cas Number : 20595-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-194
Synonym (E)-3-(2,5-Dichlorophenyl)acrylic acid, (2E)-3-(2,5-Dichlorophenyl)prop-2-enoic acid
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37188

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37188
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart