APO455826770 Apollo Scientific Ltd

6-(Dimethylamino)nicotinonitrile

  • MDL number MFCD08272099
  • Cas Number : 154924-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-68
Synonym 6-(Dimethylamino)pyridine-3-carbonitrile
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37199

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37199
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart