APO455826773 Apollo Scientific Ltd

Dimethyl 1H-pyrazole-3,5-dicarboxylate

  • MDL number MFCD00975602
  • Cas Number : 4077-76-3
  • Molecular Weight : 184.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-160
Synonym 3,5-Bis(methoxycarbonyl)-1H-pyrazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/dimethyl-1h-pyrazole-35-dicarboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/dimethyl-1h-pyrazole-35-dicarboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart