APO455826779 Apollo Scientific Ltd

Methyl 2-amino-1,3-thiazole-4-carboxylate

  • MDL number MFCD00622441
  • Cas Number : 118452-04-3
  • Molecular Weight : 158.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 171-176
Iupac Name methyl 2-amino-1,3-thiazole-4-carboxylate
Inchi Code 1S/C5H6N2O2S/c1-9-4(8)3-2-10-5(6)7-3/h2H,1H3,(H2,6,7)
Inchi Key WYVZZWKIKAKUKV-UHFFFAOYSA-N

Description

Synonym 2-Amino-4-(methoxycarbonyl)-1,3-thiazole
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 171-176
Product Link https://store.apolloscientific.co.uk/product/methyl-2-amino-13-thiazole-4-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-amino-13-thiazole-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart