APO455826785 Apollo Scientific Ltd

N-Methoxy-N-methylthiophene-2-carboxamide

  • MDL number MFCD03643923
  • Cas Number : 229970-94-9
  • Molecular Weight : 171.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methoxy-N-methyl-2-thiophenecarboxamide
Inchi Code 1S/C7H9NO2S/c1-8(10-2)7(9)6-4-3-5-11-6/h3-5H,1-2H3
Inchi Key DGMGILGPRYQSOX-UHFFFAOYSA-N

Description

Synonym 2-[Methoxy(methyl)carbamoyl]thiophene
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-methoxy-n-methylthiophene-2-carboxamide

Safety Information

signalword Warning
pcodes P280;P304+P340;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/n-methoxy-n-methylthiophene-2-carboxamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart