APO455826788 Apollo Scientific Ltd

6-(Methylamino)nicotinonitrile

  • MDL number MFCD08689756
  • Cas Number : 261715-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-133
Synonym 6-(Methylamino)pyridine-3-carbonitrile
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-methylaminonicotinonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-methylaminonicotinonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart