APO455826790 Apollo Scientific Ltd

Methyl 3-(2-bromoethoxy)benzoate

  • MDL number MFCD06204041
  • Cas Number : 59516-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 110/0.038mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37258

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37258
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart