APO455826792 Apollo Scientific Ltd

5-(Chloromethyl)uracil

  • MDL number MFCD00218445
  • Cas Number : 3590-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >350
Synonym 5-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione, 5-(Chloromethyl)-2,4-dioxo-1,2,3,4-tetrahydropyrimidine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13829

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13829
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart