APO455826793 Apollo Scientific Ltd

Methyl 5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate

  • MDL number MFCD01927210
  • Cas Number : 353269-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-62
Synonym 5,6-Dihydro-2-(methoxycarbonyl)-4H-cyclopenta[b]thiophene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37270

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37270
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart