APO455826797 Apollo Scientific Ltd

2,3-Dihydro-3-(4-nitrophenyl)-2-thioxoquinazolin-4(1H)-one

  • MDL number MFCD02916354
  • Cas Number : 72176-80-8
  • Molecular Weight : 299.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 319-320
Synonym 2,3-Dihydro-3-(4-nitrophenyl)-4-oxo-2-thioxo-1H-quinazoline, 4-(4-Oxo-2-thioxo-1,4-dihydroquinazolin-3(2H)-yl)nitrobenzene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-dihydro-3-4-nitrophenyl-2-thioxoquinazolin-41h-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-dihydro-3-4-nitrophenyl-2-thioxoquinazolin-41h-one
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart