APO455826798 Apollo Scientific Ltd

4-(2-Oxoimidazolidin-1-yl)benzonitrile

  • MDL number MFCD08689735
  • Cas Number : 144655-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-181
Synonym 1-(4-Cyanophenyl)-2-oxoimidazolidine, 1-(4-Cyanophenyl)imidazolidin-2-one
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37299

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37299
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart