APO455826801 Apollo Scientific Ltd

5-Amino-6-bromopyridine-2-carbonitrile

  • MDL number MFCD09027188
  • Cas Number : 849353-22-6
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172-174
Synonym 5-Amino-6-bromopicolinonitrile, 3-Amino-2-bromo-6-cyanopyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-6-bromopyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-6-bromopyridine-2-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart