APO455826803 Apollo Scientific Ltd

Methyl 2-bromo-1,3-thiazole-4-carboxylate

  • MDL number MFCD06659908
  • Cas Number : 170235-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-122
Synonym 2-Bromo-4-(methoxycarbonyl)-1,3-thiazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29411

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29411
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart