APO455826804 Apollo Scientific Ltd

4,6-Dichloronicotinic acid

  • MDL number MFCD00234146
  • Cas Number : 73027-79-9
  • Molecular Weight : 192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-156
Synonym 2,4-Dichloropyridine-5-carboxylic acid, 4,6-Dichloropyridine-3-carboxylic acid
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/46-dichloronicotinic-acid

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/46-dichloronicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart