APO455826811 Apollo Scientific Ltd

4-Bromo-3-(hydroxymethyl)-1-methyl-1H-pyrazole

  • MDL number MFCD06202888
  • Cas Number : 915707-65-2
  • Molecular Weight : 191.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-95
Synonym (4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-hydroxymethyl-1-methyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-hydroxymethyl-1-methyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart