APO455826812 Apollo Scientific Ltd

5-Methylisoxazole-3-carbonitrile

  • MDL number MFCD09064940
  • Cas Number : 57351-99-2
  • Molecular Weight : 108.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cyano-5-methylisoxazole
Purity 97
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methylisoxazole-3-carbonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methylisoxazole-3-carbonitrile
pictograms GHS06
hstatements H301+H311;H330
Availability details for
Quantity
  • Availability
  • Add to Cart