APO455826813 Apollo Scientific Ltd

1-Methyl-1H-pyrazole-3-carbonitrile

  • MDL number MFCD00466343
  • Cas Number : 79080-39-0
  • Molecular Weight : 107.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 41.5-45
Iupac Name 1-methyl-1H-pyrazole-3-carbonitrile
Inchi Code 1S/C5H5N3/c1-8-3-2-5(4-6)7-8/h2-3H,1H3
Inchi Key UOQLZFAAFSUXFE-UHFFFAOYSA-N

Description

Synonym 3-Cyano-1-methyl-1H-pyrazole
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 41.5-45
Product Link https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrazole-3-carbonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrazole-3-carbonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart