APO455826815 Apollo Scientific Ltd

4-[3-(Dimethylamino)propoxy]benzeneboronic acid, pinacol ester

  • MDL number MFCD09702395
  • Cas Number : 627899-90-5
  • Molecular Weight : 305.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-{4-[3-(Dimethylamino)propoxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N,N-Dimethyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propylamine
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-3-dimethylaminopropoxybenzeneboronic-acid-pinacol-ester

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-3-dimethylaminopropoxybenzeneboronic-acid-pinacol-ester
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart