APO455826819 Apollo Scientific Ltd

2-Chloro-3,5-dimethylpyrazine

  • MDL number MFCD00126945
  • Cas Number : 38557-72-1
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-3,5-dimethyl-1,4-diazine
Purity 97
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-35-dimethylpyrazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-35-dimethylpyrazine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart