APO455826823 Apollo Scientific Ltd

2-(Morpholin-4-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09702410
  • Cas Number : 906353-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-177
Synonym 4-Carboxy-2-(morpholin-4-yl)-1,3-thiazole, 4-(4-Carboxy-1,3-thiazol-2-yl)morpholine
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37922

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37922
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart