APO455826824 Apollo Scientific Ltd

4-Bromo-2-oxindole

  • MDL number MFCD06659911
  • Cas Number : 99365-48-7
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 202-208
Iupac Name 4-bromo-1,3-dihydro-2H-indol-2-one
Inchi Code 1S/C8H6BrNO/c9-6-2-1-3-7-5(6)4-8(11)10-7/h1-4,10-11H
Inchi Key YMLSQZJPMWLCQL-UHFFFAOYSA-N

Description

Synonym 4-Bromo-1,3-dihydro-2H-indol-2-one
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 202-208
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-oxindole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-oxindole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart