APO455826828 Apollo Scientific Ltd

4-(Aminomethyl)-1-methyl-1H-imidazole

  • MDL number MFCD06411536
  • Cas Number : 486414-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1-Methyl-1H-imidazol-4-yl)methylamine
Purity 97
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29314

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=29314
Availability details for
Quantity
  • Availability
  • Add to Cart