APO455826835 Apollo Scientific Ltd

1-(Piperidin-4-ylcarbonyl)azepane

  • MDL number MFCD03372514
  • Cas Number : 86542-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-46
Synonym 1-Azepanyl(piperidin-4-yl)methanone
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29325

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29325
pictograms GHS05
hstatements H314;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart