APO455826836 Apollo Scientific Ltd

5-Amino-1,3-benzothiazole

  • MDL number MFCD04115282
  • Cas Number : 1123-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-78
Synonym 1,3-Benzothiazol-5-amine, 5-Aminobenzo[d]thiazole
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-amino-13-benzothiazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-13-benzothiazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart