APO455826838 Apollo Scientific Ltd

4-(Piperazin-4-yl)benzonitrile, N1-BOC protected

  • MDL number MFCD03791198
  • Cas Number : 186650-98-6
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-125
Synonym tert-Butyl 4-(4-cyanophenyl)piperazine-1-carboxylate
Purity 97
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-piperazin-4-ylbenzonitrile-n1-boc-protected

Safety Information

signalword Warning
pcodes P271;P280;P261
MSDS https://store.apolloscientific.co.uk/product/4-piperazin-4-ylbenzonitrile-n1-boc-protected
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart