APO455826839 Apollo Scientific Ltd

Indoline-2-carboxylic acid, N-BOC protected

  • MDL number MFCD02682411
  • Cas Number : 137088-51-8
  • Molecular Weight : 263.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-122
Synonym 1-(tert-Butoxycarbonyl)-2,3-dihydro-1H-indole-2-carboxylic acid, 1-(tert-Butoxycarbonyl)-2-carboxyindoline
Purity 90
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/indoline-2-carboxylic-acid-n-boc-protected

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/indoline-2-carboxylic-acid-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart