APO455826840 Apollo Scientific Ltd

8-Azabicyclo[3.2.1]octan-3-one, N-BOC protected

  • MDL number MFCD00673779
  • Cas Number : 185099-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-74
Synonym tert-Butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, N-(tert-Butoxycarbonyl)nortropinone
Purity 98
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/8-azabicyclo321octan-3-one-n-boc-protected

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/8-azabicyclo321octan-3-one-n-boc-protected
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart