APO455826844 Apollo Scientific Ltd

Dibenzo[b,d]furan-2-carboxaldehyde

  • MDL number MFCD00136272
  • Cas Number : 5397-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-74
Synonym 2-Formyldibenzo[b,d]furan
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/dibenzobdfuran-2-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/dibenzobdfuran-2-carboxaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart