APO455826862 Apollo Scientific Ltd

Methyl 3-amino-5-hydroxybenzoate

  • MDL number MFCD09263220
  • Cas Number : 67973-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-126
Synonym 3-Amino-5-(methoxycarbonyl)phenol, 3-Hydroxy-5-(methoxycarbonyl)aniline
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38072

Safety Information

signalword Warning
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38072
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart