APO455826867 Apollo Scientific Ltd

1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride

  • MDL number MFCD09817460
  • Cas Number : 906352-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227.5-231.5
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38120

Safety Information

signalword Danger
pcodes P232;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38120
pictograms GHS05;GHS07
hstatements H302;H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart