APO455826880 Apollo Scientific Ltd

2-(4-Methoxyphenyl)pyrrolidine

  • MDL number MFCD01631836
  • Cas Number : 74190-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Pyrrolidin-2-yl)anisole, 1-Methoxy-4-(pyrrolidin-2-yl)benzene
Purity 95
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27761

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=27761
Availability details for
Quantity
  • Availability
  • Add to Cart