APO455826887 Apollo Scientific Ltd

2-Isobutylpyrrolidine hydrochloride

  • MDL number MFCD03840008
  • Cas Number : 1184994-37-3
  • Molecular Weight : 163.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(2-Methylprop-1-yl)pyrrolidine hydrochloride, 2-Isobutyltetrahydropyrrole hydrochloride
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-isobutylpyrrolidine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-isobutylpyrrolidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart