APO455826889 Apollo Scientific Ltd

3-(Chloromethyl)-5-(3,5-dimethylisoxazol-4-yl)-1,2,4-oxadiazole

  • MDL number MFCD00052098
  • Cas Number : 175205-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13793

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13793
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart