APO455826903 Apollo Scientific Ltd

3-(4-Chlorophenoxy)benzenesulphonyl chloride

  • MDL number MFCD02089485
  • Cas Number : 501697-55-8
  • Molecular Weight : 303.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-chlorophenoxy)benzenesulfonyl chloride
Inchi Code 1S/C12H8Cl2O3S/c13-9-4-6-10(7-5-9)17-11-2-1-3-12(8-11)18(14,15)16/h1-8H
Inchi Key YLAIBEFXDCITAC-UHFFFAOYSA-N

Description

Synonym 4-Chloro-3'-(chlorosulphonyl)diphenyl ether
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-4-chlorophenoxybenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-4-chlorophenoxybenzenesulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart