APO455826905 Apollo Scientific Ltd

3-(2-Methoxyphenoxy)benzenesulphonyl chloride

  • MDL number MFCD01631872
  • Cas Number : 521980-26-7
  • Molecular Weight : 298.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-methoxyphenoxy)benzenesulfonyl chloride
Inchi Code 1S/C13H11ClO4S/c1-17-12-7-2-3-8-13(12)18-10-5-4-6-11(9-10)19(14,15)16/h2-9H,1H3
Inchi Key OKUIQFDHDUXTFO-UHFFFAOYSA-N

Description

Synonym 3-(Chlorosulphonyl)-2'-methoxydiphenyl ether, 2-[3-(Chlorosulphonyl)phenoxy]anisole
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-2-methoxyphenoxybenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-2-methoxyphenoxybenzenesulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart