APO455826908 Apollo Scientific Ltd

3-(Pyridin-2-yl)benzaldehyde

  • MDL number MFCD02684103
  • Cas Number : 85553-53-3
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(3-Formylphenyl)pyridine
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-pyridin-2-ylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-pyridin-2-ylbenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H332
Availability details for
Quantity
  • Availability
  • Add to Cart