APO455826914 Apollo Scientific Ltd

N-Methyl-4-phenylbutylamine hydrochloride

  • MDL number MFCD08445574
  • Cas Number : 148252-36-2
  • Molecular Weight : 199.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Methylamino)-4-phenylbutane hydrochloride, [4-(Methylamino)but-1-yl]benzene hydrochloride
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-methyl-4-phenylbutylamine-hydrochloride

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-methyl-4-phenylbutylamine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart