APO455826916 Apollo Scientific Ltd

4-Phenoxypiperidine hydrochloride

  • MDL number MFCD02684106
  • Cas Number : 3413-27-2
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [(Piperidin-4-yl)oxy]benzene hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-phenoxypiperidine-hydrochloride

Safety Information

signalword Warning
pcodes P280;P302+P352;P362+P364
MSDS https://store.apolloscientific.co.uk/product/4-phenoxypiperidine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart