APO455826917 Apollo Scientific Ltd

4-Phenoxybenzylamine

  • MDL number MFCD01310836
  • Cas Number : 107622-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (4-Phenoxyphenyl)methylamine
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 138/5mm
Product Link https://store.apolloscientific.co.uk/product/4-phenoxybenzylamine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-phenoxybenzylamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart