APO455826919 Apollo Scientific Ltd

[4-(4-Methylphenoxy)phenyl]methylamine hydrochloride

  • MDL number MFCD02089411
  • Cas Number : 262862-66-8
  • Molecular Weight : 249.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(4-Methylphenoxy)benzylamine hydrochloride, 4-(Aminomethyl)-4'-methyldiphenyl ether hydrochloride
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-4-methylphenoxyphenylmethylamine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-methylphenoxyphenylmethylamine-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart