APO455826921 Apollo Scientific Ltd

[4-(4-Chlorophenoxy)phenyl]methylamine hydrochloride

  • MDL number MFCD02089410
  • Cas Number : 262862-71-5
  • Molecular Weight : 270.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(4-Chlorophenoxy)benzylamine hydrochloride, 4-(Aminomethyl)-4'-chlorodiphenyl ether hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-4-chlorophenoxyphenylmethylamine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-chlorophenoxyphenylmethylamine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart