APO455826930 Apollo Scientific Ltd

4-(Aminomethyl)-4'-methylbiphenyl hydrochloride

  • MDL number MFCD02089428
  • Cas Number : 1173246-91-7
  • Molecular Weight : 233.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (4'-Methylbiphenyl-4-yl)methylamine hydrochloride, 4-(4-Methylphenyl)benzylamine hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-aminomethyl-4-methylbiphenyl-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminomethyl-4-methylbiphenyl-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart