APO455826937 Apollo Scientific Ltd

3',4'-Dichloro-[1,1'-biphenyl]-4-carboxaldehyde

  • MDL number MFCD03002047
  • Cas Number : 50670-78-5
  • Molecular Weight : 251.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-98
Synonym 4-(3,4-Dichlorophenyl)benzaldehyde, 3',4'-Dichloro-4-formylbiphenyl
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/34-dichloro-11-biphenyl-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/34-dichloro-11-biphenyl-4-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart